Learn More
Falcon™ 96-Square Well, Angled-Bottom Plate and Lid
Ideal for both manual and robotic screening of compounds in cell-based assays
Brand: Falcon™ 353925
Code : 66
Additional Details : Weight : 0.42300kg
Description
Falcon™ 96 Square Well Angled Bottom Multiwell Insert System, with lid, sterile, 5/case, Non-TC-treated polystyrene, nonpyrogenic. The Falcon 96 Multiwell Insert System is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays.Storage: Store at room temperature, away from direct sunlight.
Specifications
Standard | |
8 x 12 | |
96 | |
Non-treated | |
5/Cs. | |
5/Cs. |
With Lid | |
Polystyrene | |
Sterile | |
Insert, feeder tray and lid | |
5/Pk. |