missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Fibronectin, Human
Promotes cellular attachment, spreading, and proliferation on tissue-culture surfaces. Corning™ Fibronectin, Human can be used as an additive to serum-free culture media.
£137.88 - £1640.00
Specifications
Content And Storage | 2° to 8°C |
---|---|
Product Type | Fibronectin |
pH | 11 |
Source | Human Plasma |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11533610
|
Corning™
356008 |
5 mg |
£461.69
5mg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10526961
|
Corning™
354008 |
1 mg |
£137.88
1mg |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11543610
|
Corning™
356009 |
25 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
The principal functions of fibronectin appear to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis.
Specifications
2° to 8°C | |
Fibronectin | |
11 | |
Human Plasma |