missing translation for 'onlineSavingsMsg'
Learn More

Corning™ FiltrEX Filter Plates

Individual filter discs eliminate liquid cross-contamination

Brand:  Corning™ 3511

Code : 66

Additional Details : Weight : 0.01123kg

 View more versions of this product

Product Code. 10002541

  • £1590.43 / Pack of 50
Estimated Shipment: 18-12-2024
to see stock.
Add to basket

Description

Description

Corning FiltrEX filter microplates meet standard ANSI/SBS footprint dimensions for microplates. The rigid side walls make the microplate ideal for automation and the wide skirt accepts bar codes. Individual filter disks are encapsulated in the microplate by a unique process that ensures 100% integrity of each well. The design of the nozzle prevents sample cross-contamination and wicking. Glass fiber filter microplates can be used for a variety of applications, such as plasmid isolation, DNA purification, PCR clean-up, or receptor/ligand binding assays. They are a cost-saving alternative to DNA prep kits. Use the low-binding hydrophilic PVDF membrane for lysate clarification, protein kinase assays, or bead- or resin-based separation assays. For additional application information, visit our Technical Resources section at www.corning.com/lifesciences.

Order Info

Sterile plates are wrapped in packs of 5; 10 packs per case. Lids are not included, but may be ordered separately.

Specifications

Specifications

ANSI, SBS1
Without Lid
8 x 12
White
Polystyrene, Glass Fiber
96
Filter
50/Cs.
50/Cs.
Standard
Hydrophilic PVDF
0.66 mm
Filtering Microplate
350 μL
Non-sterile
Ideal for automated liquid handling, centrifugation and vacuum filtration, wide skirt easily accommodates bar code labels
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
View accessory products
Special Offers

Special Offers