missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Flexible Packaging Systems: EVA Collection Bags

Meet FDA requirements as 510K Medical Device

Brand:  Corning™ 91-200-36

 View more versions of this product

Product Code. 17253574

  • £68.30 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.

Order Info

The full line of bag types include: collection containers (bottom port and face port designs); tank liners (2D and 3D); cryopreservation bags; cell expansion bags; bags for HYPERStack™ vessels; and rocker cell culture bags. Collection bags, collection containers and media totes are sold individually on a per each basis.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 L
1/Ea.
Male MPC plug, male MPC connector with 0.25 in. barb directly connected to bag, valve and female luer
EVA
1/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers