missing translation for 'onlineSavingsMsg'
Learn More

Corning™ GL45 Disposable Aseptic Transfer Cap
GREENER_CHOICEBIOPHARMA

Reduces contamination and enhancse the productivity of cell culture operations when adding or removing medium and cells

Brand:  Corning™ 3565

 View more versions of this product

Product Code. 11379464

  • £401.00 / Pack of 2
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Disposable Corning™ Spinner Flask Aseptic Transfer Caps will enhance the productivity of your cell culture operations when adding or removing medium and cells. Aseptic Transfer Caps are sterile (10-6 SAL), individually double bagged and fit the side arms on Corning™ Disposable Spinner Flasks

  • Cap is made of high density polyethylene; autoclaving is not recommended.
  • The venting port ends in a 25 mm diameter vent filter with a 0.2 μm pore PVDF membrane and a barbed fitting.
  • The liquid transfer port has an internal end with a rigid polypropylene dip tube angled to reach the bottom of the flask and on the external side a 1/8 inch ID (3.2 mm), 1/4 inch OD chemically resistant, heat sealable flexible tubing, ending in a male Luer lock (MLL) connector closed with a female Luer sealing cap.
  • Designed for both suspension and adherent cell culture applications and optimized for use with Corning microcarriers.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orange
2.12 in.
HDPE
125 in. Dip tube, 0.2μm vent, male leur lock
1/Pk.
GL 45
Sterile
5.4 cm
1 tube; Tube style: Angled
0.63 cm
500 mL Plastic Spinner Flask
2/Cs.
Aseptic transfer cap
Single-use
Filling
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.