Learn More
Corning™ Hybridization Chamber
Designed to hold 25 x 75mm microarray slides at a constant humidity during hybridization incubations
Brand:  Corning™ 2551
Product Code. 10286621
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Polycarbonate and nonsterile
  • O-ring and retaining clips assure the reusable chamber remains watertight in waterbaths and airtight in hybridization ovens
  • Wells in the base hold 10μL of water to maintain optimal interior humidity
  • Increased interior depth allows for single slide hybridizations and for user to place two arrays face-to-face and hybridize using a single labeled target
  • Used with raised-edge coverslips that are thicker or taller than standard thin coverglass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Wells in the base hold 10μL of water to maintain optimal interior humidity, increased interior depth allows for single slide hybridizations and for user to place two arrays face-to-face and hybridize using a single labeled target
Polycarbonate
Hybridization Chamber
5 Case