Learn More
Corning™ HYPERStack™ Accessories: Stack Manipulator
Accessories for use with Corning HYPERStack products
Brand:  Corning™ 11000
Code : 67
Additional Details : Weight : 58.96696kg
Product Code. 11367691
  • £22490.00 / Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • Made of stainless steel for durability
  • Accommodates both HYPERStack-36 and CellSTACK-10 vessels
  • Ergonomic design
  • Easy dials for protocol development
  • Handle two units simultaneously or one unit at a time
Specifications
Specifications
Stainless Steel
1/Cs.
Stack manipulator
Hyperstack™ 110and Hyperstack™ 36 Vessels
1/Pk.