missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CellBIND™ HYPERStack™ Culture Vessels

Closed system that allows fluid manipulations to occur through preassembled tubing sets

Brand:  Corning™ 10012

Code : 67

Additional Details : Weight : 11.88411kg

 View more versions of this product

Product Code. 11327671

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Closed System for High Yield Cell Growth

Introducing the next generation in Corning’s High Yield PERformance (HYPER) platform – the Corning HYPERStack cell culture vessel. This product line combines the best of two Corning products: the Corning CellSTACK™ culture chamber and the Corning HYPERFlask™ vessel. The utilization of the proprietary gas-permeable film technology provided in the format of the CellSTACK culture chamber allows the HYPERStack vessel to be the most efficient, scalable cell culture vessel for adherent cell culture available today.

Features and Benefits

  • More cells – 2.5x more cells per volumetric footprint
  • Closed system – no open fluid manipulations
  • Scalable product – multiple size offerings
  • Ergonomic design – easier manipulation
  • Fixed media volume – 0.2 mL/cm² fills vessel for optimal handling
  • Innovative assembly – no adhesives, low particulate
  • Less volumetric waste – less waste per growth area lowers overall costs

Surface Treatment Guide

The Corning CellBIND surface is designed to enhance cell attachment and increase cell yields. This enhanced surface treatment can often improve cell attachment of loosely adherent cells and maintain growth in reduced-serum or serum-free conditions. Alternatively, a Non-CellBIND option (named as Untreated) can support conventional cell attachment and allow for a faster and gentler harvest procedure (i.e., growing and harvesting cells for downstream assays).

For more information, please refer to the Corning™ CellBIND™ Surface Brochure in the resource tab on the product pages below.

Protocol Guides

Select HYPERStack video protocols available at www.corning.com/HYPER

Specifications

Specifications

Clear
4/Cs.
12-layer vessel
Sterile
6000 cm²
Gamma sterilized to SAL 10-6 standard
Polycarbonate
1/Pk.
No
Rectangular
4/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers