Learn More
Corning™ Hink's TNM-FH Medium
Used for low endotoxin content and better results.
Brand: Corning™ 13-100-CV
Code : NEW
Additional Details : Weight : 3.76000kg
Description
- Modified Grace's formulation containing lactalbumin hydrolysate (LAH) and ultrafiltered yeastolate
- Used in production of recombinant genes due to their fast doubling times and ability to grow in suspension
Order Info
Liquid and powdered media are packaged in PET with tamper-evident seals; shipped at ambient temperature. Certificates of Analysis for each lot are available from Mediatech upon request.
Compliance
Liquid products are tested for sterility according to FDA Guidelines for the Manufacture of In Vitro Diagnostic Products (January 1994) for sterile labeling.
Specifications
2° to 8°C | |
1 X | |
Hink's TNM-FH medium | |
Glutamine, Lactalbumin Hydrolysate, Yeastolate | |
3.3559 EU/ mL | |
6.3 | |
RUO |
Insect Cell | |
Liquid | |
Sterile | |
No Insect Hemolymph | |
Cell Culture | |
6 x 500 mL |