Learn More
Corning™ Lambda Plus 8 and 12-Channel Pipettors
GREENER_CHOICE
Designed for universal fit with all common brands of pipet tips
Brand:  Corning™ 4086
 View more versions of this product
Product Code. 11393625
  • £565.00 / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Engineered to provide the highest levels of accuracy, reproducibility, and comfort
Available in single-, eight-and twelve-channel configurations, all Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
Features
  • Factory calibrated to strict ISO 8655 standards-3-year warranty
  • Fully autoclavable and UV resistant
  • Designed for universal fit with all common brands of pipet tips
  • Adjustable tip ejector
Order Info
Accessory stand and rack available separately (Cat. Nos. 07-201-790 and -789).
Services
3-Year
TRUSTED_SUSTAINABILITY
Specifications
Specifications
±3 to ±1%
Autoclavable
12
Non-sterile
20 to 200 μL
3 Years
CE
Universal Pipette Tip
Lambda™ Plus
Pipette
Yes