Learn More
Corning™ Lambda Plus Pipetor Starter Kit
Designed for laboratories which are about to replace their old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes
Brand: Corning™ 4069
Code : NEW
Additional Details : Weight : 2.17724kg
Description
The Lambda Plus Starter Kit is designed for laboratories which are about toreplace their old set of pipettors, and for customers who want to get a newset of pipettors which cover the most commonly used volumes.
The Lambda Plus Starter Kit contains: four Lambda Plus Single-Channelpipettors conveniently packaged with useful accessories (manual, calibrationkey, color-coded rings, tube of piston grease and identification stickers).
- Lambda Plus Single-Channel pipettor:
- 0.5 to 10 μL (Cat. No. 4071)
- 2 to 20 μL (Cat. No. 4072)
- 20 to 200 μL (Cat. No. 4074)
- 100 to 1000 μL (Cat. No. 4075) - Universal carousel rack for six single-channel pipettors
- Corning DeckWorks™ Pipet Tips:
- 10 μL (Cat. No. 4135)
- 200 μL (Cat. No. 4138)
- 1000 μL (Cat. No. 4140) - Free Gift - 1 Corning insulated lunch bag
Specifications
1 | |
Lambda Plus | |
Starter Kit | |
Four single channel pipette, manual, calibration key, color coded rings, tube of piston grease, identification stickers |