Learn More
Corning™ Laminin/Entactin High Concentration Extracellular Matrix Protein
Provides a highly defined 3D culture environment for cell differentiation assays
Marque: Corning™ 354259
Code : 67
Informations supplémentaires : Weight : 0.02650kg
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Laminin is a major component of basement membranes. It has numerous biological activities including promotion of cell adhesion, migration, chemotaxis, growth, and differentiation, including neurite outgrowth.
Spécification
Bioassay, ELISA Standard | |
Laminin/Entactin | |
>95% by SDS-PAGE | |
Engelbreth Holm Swarm (EHS) Mouse tumor | |
Mouse |
High concentration Laminin/Entactin complex frozen in Dulbecco's PBS | |
SDS-PAGE | |
10.5 mg | |
-70°C | |
Liquid |
For research use only.