Learn More
Corning™ LSE™ Digital Dry Bath Accessory, Single Blocks
Choice of configurations for use with LSE Digital Dry Bath Heater
£102.59 - £114.11
Product Code | Brand | No. of Places | Tube Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Places | Tube Size | Price | Quantity & Availability | ||||
11373815
|
Corning™
480117 |
24 | 2 mL |
£114.11
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
11383815
|
Corning™
480118 |
24 | 0.5 mL |
£109.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
11393815
|
Corning™
480119 |
24 | 1.5 mL |
£112.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
11313825
|
Corning™
480121 |
20 | 10 mm |
£102.59
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
11323825
|
Corning™
480122 |
20 | 12 mm |
£104.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
11333825
|
Corning™
480123 |
20 | 13 mm |
£102.59
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
11971891
|
Corning™
480124 |
12 | 15 mL |
£112.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
11353825
|
Corning™
480125 |
12 | 16 mm |
£109.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
11921901
|
Corning™
480129 |
5 | 50 mL |
N/A
|
N/A | ||||
11383825
|
Corning™
480128 |
6 | 25 mm |
£102.59
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
Description
The single and dual block configurations are used in the Corning LSE Digital Dry Bath Heaters.
Specifications
Stainless Steel | |
Block |