Learn More
Corning™ LSE™ Vortex Mixer Accessory, Processing Heads
Heads allow for processing of larger numbers of tubes
Brand:  Corning™ 480101
Code : 04
Additional Details : Weight : 0.18450kg
 View more versions of this product
Product Code. 11921981
  • £34.46 / Each
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
Description
Description
This is an optional head for processing larger numbers of tubes, for use with Corning™ LSETM Vortex Mixer.
Specifications
Specifications
24 x 1.5/2.0 mL tubes, 24 x 0.5 mL tubes and 32 x 0.2 mL tubes/4 tube strips
LSE Vortex Mixer