Learn More
Corning™ LSE™ Vortex Mixer Accessory, Processing Heads
Heads allow for processing of larger numbers of tubes
Brand:  Corning™ 480102
Code : 04
Additional Details : Weight : 0.21772kg
 View more versions of this product
Product Code. 11941981
  • £42.59 / Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
This is an optional head for processing larger numbers of tubes, for use with Corning™ LSETM Vortex Mixer.
Specifications
Specifications
1 Microplate, 64 x 0.2 mL tubes, 8 x 0.2 mL tube strips
LSE Vortex Mixer