missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Matrigel™ Matrix
GREENER_CHOICEBIOPHARMA

Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.

£310.00 - £1641.46

Specifications

Content And Storage -20°C
Product Type Basement membrane matrix
For Use With (Application) Cell growth and differentiation; metabolism/toxicology studies; invasion assays; in vitro and in vivo angiogenesis assays; in vivo angiogenesis studies and augmentation of tumors in immunosuppressed mice
Source Engelbreth-Holm-Swarm Mouse Tumor
View More Specs

Alert:

Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer


Products 4
Product Code Brand Quantity Format Price Quantity & Availability  
Product Code Brand Quantity Format Price Quantity & Availability  
10429212
View Documents
Corning™
356235
50 mL Matrigel Matrix LDEV-Free This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11573620
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Corning™
356234
5 mL Matrigel Matrix LDEV-Free
£310.00
5mL
Please to purchase this item. Need a web account? Register with us today!
11593620
GREENER_CHOICE
View Documents Promotion Details
Corning™
356237
10 mL Matrigel Matrix Phenol Red-Free, LDEV-Free
£540.00
10mL
Please to purchase this item. Need a web account? Register with us today!
11543550
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Corning™
354234
10 mL Matrigel Matrix LDEV-Free
£439.00
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Specifications

Specifications

-20°C
Basement membrane matrix
Cell growth and differentiation; metabolism/toxicology studies; invasion assays; in vitro and in vivo angiogenesis assays; in vivo angiogenesis studies and augmentation of tumors in immunosuppressed mice
Engelbreth-Holm-Swarm Mouse Tumor
Videos
SDS
Documents

Documents

Product Certifications