Learn More
Corning™ Matrigel™ GFR Basement Membrane Matrix

Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ GFR Basement Membrane Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
£340.00 - £570.00
Specifications
Content And Storage | -20°C |
---|---|
Product Type | GFR Basement Membrane Matrix |
For Use With (Application) | Suitable for applications that require a highly defined basement membrane preparation. |
Source | Engelbreth-Holm-Swarm Mouse Tumor |
Alert:
Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer
Product Code | Brand | Quantity | Without Additives | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Without Additives | Price | Quantity & Availability | ||||
11543620
![]() |
Corning™
356230 |
5 mL | No LDEV |
£340.00
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11523550
![]() |
Corning™
354230 |
10 mL | No LDEV |
£540.00
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11553620
![]() |
Corning™
356231 |
10 mL | No Phenol Red, No LDEV |
£570.00
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
-20°C | |
GFR Basement Membrane Matrix | |
Suitable for applications that require a highly defined basement membrane preparation. | |
Engelbreth-Holm-Swarm Mouse Tumor |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.