missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Microarray Storage Pouches

Product Code. p-2915418 Shop All Corning Products
Click to view available options
Description:
For 25 Slide Storage Box
For 5 Slide Mailer
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10275842

Brand: Corning™ 40086

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Protect printed microarrays from light, humidity, and environmental contaminants

  • Tear-resistant, foil laminated pouches with 3 x 4 in. white marking label
  • Heat-sealable
  • Nonsterile

TRUSTED_SUSTAINABILITY

Specifications

Description For 25 Slide Storage Box
Quantity 50/Cs.
Color Gray
Type Storage Pouch
For Use With (Application) Printed microarrays
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.