Learn More
Corning™ Sterile Microspatulas, rounded end
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™ Sterile Microspatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
Brand: Corning™ 3013
Description
- Corning spatulas are designed to save time and to provide contamination-free samples.
- Eliminates the recycling and resterilizing necessary with reusable spatulas.
- Available in five different configurations.
- Microspatulas available in two configurations.
- Individually packaged.
- Antistatic.
- Sterile.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
Sterile | |
Single-use | |
Molecular biology applications, lyophilized samples and contamination-free sampling | |
V-shaped scoop at one end and a flat rounded blade at the other end | |
15.2 cm |
USP Class VI, non-pyrogenic, RNase and DNase-free Certified | |
Micro Spatula | |
5.98 in. | |
Polystyrene |