missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 2.5S Nerve Growth Factor (NGF), Mouse Natural
Designed specifically for use in basic cell biology research
£180.06 - £2421.60
Specifications
Content And Storage | 2° to 8°C |
---|---|
Product Type | Growth Factor |
Formulation | Lyophilized from phosphate buffered saline, carrier free, reconstitute in serum containing medium or aqueous buffer containing BSA 2% |
For Use With (Application) | Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro |
Purity or Quality Grade | >90% by 4 to 20% Tris Glycine Gel Electrophoresis |
Description
- NGFs are isolated from mouse submaxillary glands
- 2.5S form consists of two β subunits
- 7S form consists of two α, one β and two ϒ subunits
- Products are tested for biological activity in appropriate assays, such as the ability of growth factors to stimulate proliferation or DNA synthesis
- Lymphokines and growth factors are also sterile filtered generally using a 0.2μm membrane
- Tested for the presence of bacteria, fungi and mycoplasma
Specification sheets include detailed information on quality control testing, product source, storage, stability and recommendations on proper experimental concentrations.
Order Info
Both supplied in lyophilized form
Specifications
2° to 8°C | |
Lyophilized from phosphate buffered saline, carrier free, reconstitute in serum containing medium or aqueous buffer containing BSA 2% | |
>90% by 4 to 20% Tris Glycine Gel Electrophoresis | |
Mouse Submaxillary Glands |
Growth Factor | |
Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro | |
Mouse |
Safety and Handling
missing translation for 'shelfLife' : 3 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title