missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Nu-Serum™ Growth Medium Supplement
GREENER_CHOICE

Product Code. p-90900 Shop All Corning Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10632702

Brand: Corning™ 355100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides a low-serum alternative to newborn calf, fetal bovine, and other sera used for cell culture

The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.

  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions

Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Growth Supplement
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.