missing translation for 'onlineSavingsMsg'
Learn More

Corning™ NutriStem™ hPSC XF Medium

Product Code. 15627036
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 15627036

Brand: Corning™ 40051001A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Supports long-term culture of human pluripotent stem cells

Corning NutriStem hPSC XF medium is a defined, xeno-free, serum-free medium designed to support the growth and expansion of human induced pluripotent stem (hiPS) and human embryonic stem (hES) cells in a feeder-free environment.
  • Defined, xeno-free, serum-free
  • Complete, ready-to-use
  • Superior proliferation of undifferentiated hES and hiPS cells
  • Stable pluripotency and genotype over long-term culture
  • Extensively tested and widely referenced
  • TRUSTED_SUSTAINABILITY

    Specifications

    Cell Type Undifferentiated hES and hiPS cells
    Classification Serum-free, Xeno-free, cGMP
    Form Liquid
    Sterility Sterile
    With Additives HSA
    Quantity 500 mL
    Content And Storage -25° to -10°C
    Description hPSC XF Medium
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.