missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Paddle Assembly for Reusable Spinner Flasks
For use with direct drive overhead motors and series 4510 and 4512, 8L Reusable Spinner Flasks.
£330.00 - £343.23
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10485342
|
Corning™
4515-15L |
15L Corning Reusable spinner flask with direct drive overhead motor |
£330.00
Each |
In Stock
Log in to see stock. |
|||||
10315122
|
Corning™
4515-8L |
8L Corning Reusable spinner flask with direct drive overhead motor |
£331.94
Each |
In Stock
Log in to see stock. |
|||||
10683433
|
Corning™
4515-36L |
36L Corning Reusable spinner flask with direct drive overhead motor |
£343.23
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
Description
- For series 4510, 4512, and 4514 spinner flasks when coupled to a direct drive motor.
- Paddle assemblies will couple to 100 mm and 120 mm cap assemblies.
Specifications
Paddle Assembly |