Learn More
Corning™ PET Storage Bottles, Square with Cap
Shop All Corning Products

Description
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight, break-resistant and transparent as glass. Bottles use a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
Features:
- Good for storage of media, buffers and other aqueous solutions
- Wide neck provides easier access for liquid transfer
- Screened white enamel graduations are easier to read than molded graduations
- Sterilized to SAL 10−6
- Large white marking spot for easier identification and easy opening linear tear packaging
- Individually bagged
- Validated USP Class VI, noncytotoxic and nonpyrogenic

Specifications
Specifications
Capacity | 16.9 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16.9 oz. |
Color | Orange |
Diameter (English) | 3 in. |
Diameter (English) Inner Neck | 1.4 in. |
Diameter (Metric) Inner Neck | 37 mm |
For Use With (Application) | Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage |
Material | PET |
No. per Case | 24/Cs. |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
01824001 | Certificate of Quality | 29/03/2025 | 431532 |
23122011 | Certificate of Quality | 24/06/2024 | 431532 |
20922008 | Certificate of Quality | 14/10/2023 | 431532 |
01824001 | Certificate of Compliance | 17/07/2024 | 431532 |
20922008 | Certificate of Compliance | 13/03/2023 | 431532 |
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
15Minutes
14Seconds
32
Your input is important to us. Please complete this form to provide feedback related to the content on this product.