missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PET Storage Bottles, Square with Cap
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Capacity:
16.9 oz., 500 mL
33.8 oz., 1000 mL
4.2 oz., 125 mL
8.45 oz., 250 mL
Diameter (English):
2 in.
2.3 in.
3 in.
3.6 in.
Graduations:
100 to 1000 mL
100 to 500 mL
50 to 125 mL
50 to 250 mL
Height (English):
4.19 in.
5.6 in.
6.9 in.
8.5 in.
Description
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight, break-resistant and transparent as glass. Bottles use a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
Features:
- Good for storage of media, buffers and other aqueous solutions
- Wide neck provides easier access for liquid transfer
- Screened white enamel graduations are easier to read than molded graduations
- Sterilized to SAL 10−6
- Large white marking spot for easier identification and easy opening linear tear packaging
- Individually bagged
- Validated USP Class VI, noncytotoxic and nonpyrogenic
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity | 4.2 oz., 125 mL |
Capacity (Metric) | 125 mL |
Capacity (English) | 4.2 oz. |
Color | Orange |
Diameter (English) | 2 in. |
Diameter (English) Inner Neck | 1.4 in. |
Diameter (Metric) Inner Neck | 37 mm |
For Use With (Application) | Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage |
Material | PET |
No. per Case | 24/Cs. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'