missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Polyetherimide Centrifuge Tube Cushions
GREENER_CHOICEBIOPHARMA

Tube cushion provides necessary support during centrifugation preventing large capacity tubes from collapsing

Brand:  Corning™ 431124

151.48 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.


 View more versions of this product

Product Code. 10789721

  • £193.00 / Pack of 6
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Corning™ polyetherimide centrifuge tube cushions provide the necessary support during centrifugation to prevent these large capacity tubes from collapsing. Please check our Technical Information Listing for information on the physical and chemical properties of Corning plastic centrifuge tubes as well as important suggestions for safe use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gray
Polypropylene
6
Non-sterile
16.9 oz.
Centrifuge Tube
500 mL
Corning 500 mL centrifuge tubes
1
Conical
Conical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.