Learn More
Corning™ Polypropylene Plates
Manufactured from polypropylene for optimum visibility and high chemical resistance. Corning™ Polypropylene Plates are an excellent choice for transport and long-term storage.
£318.00 - £540.00
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Color | Clear |
Material | Polypropylene |
Surface Treatment | Non-treated |
Product Code | Brand | Sterility | No. of Wells | Array | Well Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | No. of Wells | Array | Well Shape | Price | Quantity & Availability | |||
10218481
|
Corning™
3363 |
Non-sterile | 96 | 8 x 12 | Conical Bottom (V-well) |
£318.00
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
10035281
|
Corning™
3364 |
Non-sterile | 96 | 8 x 12 | Flat Bottom (F-well) |
£324.86
Pack of 100 |
In Stock
Log in to see stock. |
|||
10256141
|
Corning™
3656 |
Sterile | 384 | 16 x 24 | Round |
£540.00
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
10748592
|
Corning™
3359 |
Sterile | 96 | 8 x 12 | Round |
£374.40
Pack of 100 |
In Stock
Log in to see stock. |
|||
10584391
|
Corning™
3657 |
Non-sterile | 384 | 16 x 24 | Round |
£514.00
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
10022491
|
Corning™
3357 |
Sterile | 96 | 8 x 12 | Conical Bottom (V-well) |
£365.00
Pack of 100 |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||
10650356
|
Corning™
3365 |
Non-sterile | 96 | 8 x 12 | Round |
£318.00
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or nonsterile
Order Info
Seal with Storage Mat III (sold separately).
Specifications
Standard | |
Clear | |
Non-treated | |
100/Cs. | |
100/Cs. |
Without Lid | |
Polypropylene | |
Transport and longterm storage | |
25/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.