Learn More
Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates permit top or bottom analysis using detection microplate readers.
£1170.78 - £1265.11
Specifications
Barcode No Barcode
Format Standard
Lid Without Lid
Array 16 x 24
Binding Type Medium Binding
View More Specs
Products 2
Product Code Brand Barcode BindingProperty Color Sterility Price Quantity & Availability  
Product Code Brand Barcode BindingProperty Color Sterility Price Quantity & Availability  
10640502
View Documents
Corning™
3711
No Barcode Black Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10045281
View Documents
Corning™
3706
No Barcode White, Clear Bottom Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Molded polystyrene with opaque walls and optically clear bottoms
  • Readable from top or bottom for direct microscope viewing and CCD camera-based detectors
  • Opaque black or white walls prevent well to well crosstalk
  • Recommended working volumes of 20 to 80μL; total well volume of 112μL
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Suited for flourescent and luminescent assays
  • Plate dimensions meet proposed industry standard
Specifications
Specifications
No Barcode
Without Lid
Medium Binding
384
Non-sterile
Flat Bottom (F-well)
100/Cs.
Standard
16 x 24
Polystyrene
20 to 80 μL
Non-treated
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.