Learn More
Corning™ 384-Well Clear Polystyrene Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 384-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.
Brand: Corning™ 3702
This item is not returnable.
View return policy
Description
Specifications
Standard | |
16 x 24 | |
Polystyrene | |
20 to 80 μL | |
Non-treated | |
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions. | |
25/Pk. |
Without Lid | |
Clear | |
384 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |