missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Premium Fetal Bovine Serum

FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.

Brand:  Corning™ 35-015-CV

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15383681

  • £1040.00 / 500mL
In Stock
to see stock.

Description

Description

  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Premium FBS is sourced from U.S. herds
Specifications

Specifications

cGMP
-25° to -15°C
500 mL
Triple-filtered, 0.1 μm
Premium
Used as a supplement to cell culture media to help sustain cell growth and metabolism. This product is for research use only.
<30 mg/dL
7.8 ±0.8
USDA Inspected
Caramel/Amber
Fetal Bovine Serum
Sterile
Bovine Fetus
≤20EU/ mL
No
315 ± 35mOsm/kg
30 to 50g/L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers