missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Primaria™ Cell Culture Multiwell™ Plates
GREENER_CHOICE

Product Code. 10288830 Shop All Corning Products
Click to view available options
Well Count:
24
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10288830

Brand: Corning™ 353847

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Features a surface treatment that is a permanent, stable modification of the polystyrene surface incorporating a mixture of anionic and cationic functional groups. Corning™ Primaria™ Cell Culture Multiwell™ Plates feature a flat bottom with low evaporation lid.

  • Unique nitrogen-containing tissue culture surface chemistry
  • Improves attachment, spreading, and growth for many primary cells or cell lines
  • Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/mL
TRUSTED_SUSTAINABILITY

Specifications

Array 4 x 6
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 1 mL (Well)
Color Clear
Culture Area 2 cm2
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Primaria
Type Cell Culture Plate
Volume (Metric) Working 1.5 to 2.2 mL
Well Count 24
For Use With (Application) Suitable for growing a variety of primary cells and cell lines that grow poorly on conventional, negatively charged, standard tissue culture surfaces
Includes Lid
No. per Case 50/Cs.
No. per Pack 1/Pk.
Packaging Individually Wrapped
Quantity 50/Cs.
Length (Metric) 127.63 mm
Width (Metric) 85.47 mm
Height (Metric) 21.43 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.