Learn More
Corning™ Primaria™ Tissue Culture Flasks
Improves attachment, spreading and growth for many primary cells or cell lines. Corning™ Primaria™ Tissue Culture Flasks are suitable for growing cells that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes.
£198.25 - £396.51
Specifications
Closure Color | Blue |
---|---|
Closure Material | Phenolic |
Material | Polystyrene |
Shape | Rectangular |
Sterility | Sterile |
Product Code | Brand | Capacity (Metric) | Closure Type | Culture Area | Neck Style | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Type | Culture Area | Neck Style | Price | Quantity & Availability | ||
10196070
|
Corning™
353824 |
250 mL | Plug Seal | 75 cm2 | Straight |
£384.38
Pack of 100 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
10469222
|
Corning™
353810 |
250 mL | Vented | 75 cm2 | Straight |
£384.38
Pack of 100 |
Estimated Shipment: 06-12-2024
Log in to see stock. |
||
10556932
|
Corning™
353813 |
50 mL | Plug Seal | 25 cm2 | Canted |
£396.51
Pack of 200 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
10613672
|
Corning™
353808 |
50 mL | Vented | 25 cm2 | Canted |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Unique nitrogen-containing tissue culture surface chemistry
- Improves attachment, spreading, and growth for many primary cells or cell lines
- Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
- Stable, permanent surface modification
- Optically clear
- No special storage required
- Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
- Packaged in red color-coded, peel-open, medical-style packages
- Sterilized by gamma irradiation
- Nonpyrogenic tested to less than 0.1 EU/mL
Order Info
Smaller flasks packed 20 per sleeve in cases of 200 (08-772-45 in cases of 100); larger flasks packed five per sleeve in cases of 100
Specifications
Blue | |
Polystyrene | |
Sterile |
Phenolic | |
Rectangular | |
Rectangular |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.