Learn More
Corning™ PTFE-Coated Magnetic Stir Bars
GREENER_CHOICE
For use in Corning Hotplate Stirrers
Brand:  Corning™ 409000
 View more versions of this product
Product Code. 10626953
  • £45.85 / Each
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning magnetic stir bars are polytetrafluorethylene (PTFE)-coated for increased chemical ­resistance. 
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.75 in.
3 in.
19 mm
Magnet
Stir Bar
PC-611 hotplate stirrers
PTFE
76.2 mm
1