missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PureCoat™ Amine Flasks
Positively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays
£58.69 - £650.00
Specifications
Closure Material | Polystyrene |
---|---|
Material | Polystyrene |
For Use With (Application) | For preparative cell culture, cell-based and HTS screening assays |
No. per Pack | 5 Pk. |
Shape | Rectangular |
Product Code | Brand | Capacity (Metric) | No. per Case | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | No. per Case | Price | Quantity & Availability | ||||
11947151
|
Corning™
356728 |
750 mL | 40/Cs. |
£650.00
Pack of 40 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
10689318
|
Corning™
354726 |
250 mL | 5/Cs. |
£58.69
Pack of 5 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
10868230
|
Corning™
356726 |
250 mL | 50/Cs. |
£405.91
Pack of 50 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
13436839
|
Corning™
354728 |
750 mL | 5/Cs. |
£88.04
Pack of 5 |
In Stock
Log in to see stock. |
||||
Description
Corning™ PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.Specifications
Polystyrene | |
For preparative cell culture, cell-based and HTS screening assays | |
Rectangular | |
Sterile |
Polystyrene | |
5 Pk. | |
Amine | |
Vented |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.