Learn More
Corning™ PureCoat™ Carboxyl Multiwell Plates
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Brand: Corning™ 356775
Code : 66
Additional Details : Weight : 3.32000kg
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
4 x 6 | |
Yes | |
Flat | |
Rectangular | |
Multi-well Cell Culture Plate | |
Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested | |
50/Cs. | |
50/Cs. | |
85.47 mm |
ANSI, SBS | |
Polystyrene | |
Polystyrene | |
Sterile | |
24 | |
Lid | |
5/Pk. | |
127.63 mm | |
21.43 mm |