missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PureCoat™ Carboxyl Multiwell Plates

Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.

Brand:  Corning™ 354775

Code : 67

Additional Details : Weight : 0.46000kg

Product Code. 11593580

  • £42.65 / Pack of 5
Estimated Shipment: 26-11-2024
to see stock.
Add to basket

Description

Description

Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.

Specifications

Specifications

4 x 6
Yes
Flat
Rectangular
Multi-well Cell Culture Plate
Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested
5/Cs.
5/Cs.
85.47 mm
ANSI, SBS
Polystyrene
Polystyrene
Sterile
24
Lid
5/Pk.
127.63 mm
21.43 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers