Learn More
Corning™ PureCoat™ E cm Mimetic Multi-Flask
Features multi-layers with vented cap
£780.00 - £1085.00
Specifications
Neck Style | Straight |
---|---|
No. per Pack | 1/Pk. |
Shape | Rectangular |
Sterility | Sterile |
Bottom Shape | Rectangular |
Product Code | Brand | Description | Surface Treatment | Culture Area | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Surface Treatment | Culture Area | Price | Quantity & Availability | |||
15319830
|
Corning™
356244 |
Individually packed in foil bag | Fibronectin Peptide | 525 cm2 |
£830.00
Pack of 8 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||
15329830
|
Corning™
356245 |
Individually packed in foil bag | Fibronectin Peptide | 875 cm2 |
£1085.00
Pack of 6 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||
15349830
|
Corning™
356274 |
2 per pack x 4 | Collagen I Peptide | 525 cm2 |
£780.00
Pack of 8 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||
15359830
|
Corning™
356275 |
1 per pack x 6 | Collagen I Peptide | 875 cm2 |
£965.00
Pack of 6 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||
Description
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Compliance
cGMP manufacturing compliant (ISO 9001:2008, ISO 13485:2012)
Specifications
Straight | |
Rectangular | |
Rectangular |
1/Pk. | |
Sterile | |
Vented |