Learn More
Corning™ Fetal Bovine Serum, Regular (Heat Inactivated)
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
Brand: Corning™ 35-011-CV
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Sera is aseptically collected and processed in a cGMP facility
- Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
- Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
- Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
- Regular FBS is sourced from herds in USDA approved countries (ex: Mexico, Honduras, Costa Rica, Nicaragua, etc.)