missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Reusable Plastic Reagent Bottles with GL-45 PP Screw Cap
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
Description
These bottles are manufactured from translucent polypropylene. They are wide-mouth and supplied with PP screw top caps.
Specifications
Specifications
Capacity (Metric) | 2 L |
Color | Translucent |
Material | Polypropylene |
No. per Case | 6/Cs. |
No. per Pack | 1/Pk. |
Sterility | Non-sterile |
Shape | Cylindrical |
Height (Metric) | 24.3 cm |
Diameter (Metric) | 13.1 cm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'