missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Reusable Polypropylene Low-Form Beakers, Graduated

Translucent polypropylene beakers printed with blue graduations.

£27.60 - £126.00

Specifications

Color Translucent
Material Polypropylene
For Use With (Application) For High Temperature and Chemical Resistance
Temperature (English) Max. Operating 68°F
Temperature (Metric) Max. Operating 20°C
View More Specs

Products 11
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11327841
View Documents
Corning™
1000P-25
25 mL
£56.70
Pack of 24
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
11377841
View Documents
Corning™
1000P-50
50 mL
£31.58
Pack of 12
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
11397831
View Documents
Corning™
1000P-100
100 mL
£32.50
Pack of 12
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
11307841
View Documents
Corning™
1000P-150
150 mL
£40.05
Pack of 12
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
11337841
View Documents
Corning™
1000P-250
250 mL
£27.60
Pack of 6
In Stock
to see stock.
Edge
Add to basket
 
11367841
View Documents
Corning™
1000P-400
400 mL
£34.00
Pack of 6
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
11397841
View Documents
Corning™
1000P-600
600 mL
£44.65
Pack of 6
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
11317841
View Documents
Corning™
1000P-1L
1 L
£56.40
Pack of 6
In Stock
to see stock.
Edge
Add to basket
 
11347841
View Documents
Corning™
1000P-2L
2000 mL
£123.00
Pack of 6
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
11357841
View Documents
Corning™
1000P-3L
3000 mL
£88.15
Pack of 4
In Stock
to see stock.
Add to basket
 
11387841
View Documents
Corning™
1000P-5L
5000 mL
£126.00
Pack of 4
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Translucent polypropylene beakers printed with blue graduations.

Specifications

Specifications

Translucent
For High Temperature and Chemical Resistance
20°C
Autoclavable
Yes
Blue Graduated Scale
Polypropylene
68°F
Beaker
Reusable
DIN, ASTM Class A and B
Griffin
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.