Learn More
Corning™ Sealing Mat for 384-Well Polypropylene Storage Plates
Ideal for use in long-term, low temperature compound storage
Brand:  Corning™ 3341
Product Code. 10572383
  • £479.85 / Pack of 50
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Precise fit for 384 well Deep Well Polypropylene Storage Block (Item #’s 3342 and 3347)
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g. DMSO, ethanol, methanol)
  • Firm seal prevents evaporation and gas exchange
  • Ideal for use in long-term, low temperature compound storage
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNase and RNase-free
Non-sterile
50/Cs.
Sealing Mat
384-well Deep Well Storage Block
Polypropylene
384
10/Pk.
V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products