Learn More
Corning™ Sealing Mat for 96-Well Polypropylene Microplates
Firm seal prevents evaporation and gas exchange
Brand: Corning™ 3346
This item is not returnable.
View return policy
Description
- Precise fit for 96 well Expanded Volume Polypropylene Microplate (Item #’s 3343 and 3344)
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g. DMSO, ethanol, methanol)
- Firm seal prevents evaporation and gas exchange
- Ideal for use in long-term, low temperature compound storage
Specifications
Non-sterile | |
50/Cs. | |
Sealing Mat |
96 | |
10/Pk. | |
96-well Polypropylene Plate |