Learn More
Corning™ Sealing Mat for 96-well Polypropylene Plates
For use with polypropylene plates (07-200-700, -701)
Brand: Corning™ 3083
Code : 66
Additional Details : Weight : 0.86182kg
Description
- Manufactured from EVA (ethyl vinyl acetate) and polymer is compatible with DMSO
- DNase- and RNase-free
- Can be applied manually or with our Storage Mat Applicator
- Nonsterile
Specifications
DNase and RNase-free | |
Non-sterile | |
50/Cs. | |
Sealing Mat | |
96-well Polypropylene Plate |
EVA, Polymer | |
96 | |
1/Pk. | |
Square |