Learn More
Corning™ Selenous Acid (Sodium Salt)
Shown to promote proliferation of MDCK cells in serum-free medium
Brand: Corning™ 354201
Code : 67
Additional Details : Weight : 0.00010kg Transport : UN number : 2630 Chem class : 6.1 Pack group : I
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Shown to promote proliferation of MDCK cells in serum-free medium
Specifications
2° to 8°C | |
Selenous Acid | |
1000 mg |