missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Smooth 1536-Well Multiple Well Plates
GREENER_CHOICE

Ensure precision liquid handling

£862.76

Specifications

Format Standard
Lid Without Lid
Array 32 x 48
For Use With (Equipment) Automated dispensers including the PerkinElmer FlexDrop™
Material Polystyrene
View More Specs

Products 2
Product Code Brand For Use With (Application) Color Sterility Surface Treatment Price Quantity & Availability  
Product Code Brand For Use With (Application) Color Sterility Surface Treatment Price Quantity & Availability  
12751935
GREENER_CHOICE
View Documents
Corning™
For Kalypsys™ systems, ensure precision liquid handling Black Non-sterile Non-treated
£862.76
Pack of 50
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
12761935
GREENER_CHOICE
View Documents
Corning™
7247
For Kalypsys™ systems, ensure precision liquid handling White Non-sterile Non-treated
£862.76
Pack of 50
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
Description

Description

Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.

  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Specifications

Specifications

Standard
32 x 48
Polystyrene
8 μL
Non-treated
For Kalypsys™ systems, ensure precision liquid handling
10/Pk.
Without Lid
Automated dispensers including the PerkinElmer FlexDrop™
1536
Non-sterile
Flat Bottom (F-well)
50/Cs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.