missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Square Polycarbonate Storage Bottles with 45 mm Cap
BIOPHARMA

Product Code. 10160213
Click to view available options
Capacity (Metric):
150 mL
250 mL
500 mL
1 L

Includes

Linerless, one-piece, autoclavable orange GL45 threaded polypropylene plug-seal caps

Space-Saving square shape requires less space (13 to 20%) on the shelf or in the autoclave. Corning™ Square Polycarbonate Storage Bottles are ideal for mixing, sampling and storage.

Corning™ square polycarbonate storage bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage.


Caution: These square polycarbonate bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

  • Strong polycarbonate bottles are more break resistant than other glass or plastic bottles.

  • Use a linerless, 45mm one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.

  • Screened white enamel graduations are easier to see than molded graduations

  • Sterilized by gamma radiation

  • Large white marking spot for easier identification

  • Bottles can be autoclaved once at 121oC and 15psi. Repeated autoclaving weakens polycarbonate and is not recommended.

  • Optional reusable caps with silicone septa (Corning Cat. Nos. 1395-45HTSC, 1395-45SS, 1395-45TS) for syringe sampling or introduction of reagents are available for these caps.

  • Note: Most GL45 threaded caps designed for glass storage bottles use a different sealing design and will not give a secure seal if used on these plastic bottles unless they have a rubber liner.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Capacity 7.5 oz., 250 mL
    Capacity (Metric) 250 mL
    Capacity (English) 7.5 oz.
    Material Polycarbonate
    No. per Case 24/Cs.
    No. per Pack 12/Pk.
    Sterility Sterile
    Closure Color Orange
    Graduations 50 to 150 mL
    Autoclavable Autoclavable
    Shape Square
    Closure Material Polypropylene
    Closure Size GL 45
    Closure Type Screw Cap
    Show More Show Less
    missing translation for 'provideContentCorrection'

    missing translation for 'productTitle'