Learn More
Corning™ Sterile Spatulas, Flat end



Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
Brand: Corning™ 3007
Description
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
- Corning spatulas are designed to save time and to provide contamination-free samples.
- Eliminates the recycling and resterilizing necessary with reusable spatulas.
- Available in five different configurations.
- Microspatulas available in two configurations.
- Individually packaged.
- Antistatic.
- Sterile.
- RNase-/DNase-free.
- Nonpyrogenic.

Specifications
Sterile | |
USP Class VI, non-pyrogenic, RNase and DNase-free Certified | |
Spatula | |
9.75 in. | |
Polystyrene |
Flat End, Round End | |
Single-use | |
Molecular biology applications, lyophilized samples and contamination-free sampling | |
Flat end blade at one end and a larger round spoon at the other end | |
24.76 cm |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.