Learn More
Corning™ Clear Polystyrene High Bind Stripwell™ Strips, without Frame or Lid, Non-sterile
Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Strips provide a superior solution for in vitro diagnostic assays.
Brand:  Corning™ 2580
Code : 67
Additional Details : Weight : 2.99371kg
Product Code. 10226771
  • £430.71 / Pack of 800
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
Description
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Specifications
Specifications
Strip Plate
8 x 12
Clear
96
Non-sterile
360 μL
In-vitro diagnostic assays
200/Pk.
Without Lid
High Bind
Polystyrene
75 to 200 μL
High Binding
Round, Flat Bottom (F-well)
800/Cs.
800/Cs.