missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates
Manufactured from ultrathin polycarbonate for fast, consistent heat transfer. Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.
£150.34
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Color | Clear |
Material | Polycarbonate |
Product Code | Brand | For Use With (Application) | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | For Use With (Equipment) | Price | Quantity & Availability | ||||
10777231
|
Corning™
6509 |
PCR | Applied Biosystems GeneAmp™ PCR System 9600b, Barnstead Thermolyne Amplitron II™, Techne™ Cyclogene, and Gene E with 96 x 0.2 mL block |
£150.34
Pack of 25 |
In Stock
Log in to see stock. |
||||
10130961
|
Corning™
6511 |
PCR | MJ Research PTC-100-96V, PTC-200 DNA EngineTM, Biometra Uno - ThermoblockerTM,Coy Corporation Temp Cycler II, Corbett Research FTS-960, Hybaid OmniGene withMicroblock, Quatro BioSystems T-C-40 |
£150.34
Pack of 25 |
In Stock
Log in to see stock. |
||||
Description
- Low protein-binding surface prevents loss of sample
- For oil-free operation, use polyethylene or aluminum sealing tape in conjunction with heated-lid thermal cyclers; also accommodates silicone reusable sealing mat
- Withstands temperatures to 135°C without warping
- Various well shapes fit popular thermal cyclers
- Surface of DNA-BIND™ plate allows covalent attachment of ssDNA at 5« and 3« end
- Nonsterile
Certifications
Polypropylene plates are certified DNase- and RNase-free.
Specifications
Standard | |
8 x 12 | |
Polycarbonate | |
Non-sterile | |
PCR | |
1/Pk. |
Without Lid | |
Clear | |
96 | |
Round | |
25/Cs. | |
Clear |