missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Coronavirus HCoV-EMC/2012 Proteome Microarray

Coronavirus HCoV-EMC/2012 Proteome Microarray with 3818 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.035.001

Additional Details : Weight : 0.02200kg

Product Code. 16397768

  • £482.30 / Each
Estimated Shipment: 15-10-2024
to see stock.
Add to basket

Description

Description

  • Microarray Content: Full proteome of human coronavirus HCoV-EMC/2012: replicase polyprotein 1ab, S protein, ORF3 protein, ORF4a protein, ORF4b protein, ORF5 protein, E protein, M protein, N protein, ORF8b protein
  • Peptide Length/Overlap: 15 aa/ 12 aa (replicase polyprotein 1ab) and 15aa/ 13aa (S, E, M, N proteins, ORF proteins 3, 4a, 4b, 5, 8b)
  • Number of Peptides: 3818 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEPperSlide™ PEGMA/PMMA
Antibody validation, epitope mapping, serum screening, biomarker discovery
Microscopic Glass
Individual barcode for each microarray
1 Slide
1 mm
Transparent
75.4 x 25 mm
High-density Peptide Microarray
Non-sterile
1 array copy per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers